Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205121.1 | NP_990452.1 | 2 - 3 | 345 | 104 | |
| XM_015288196.2 | XP_015143682.1 | 3 - 4 | 555 | 104 | ||
| XM_015288197.2 | XP_015143683.1 | 2 - 3 | 395 | 104 | ||
| XM_015288198.2 | XP_015143684.1 | 2 - 3 | 1330 | 104 | ||
| GenBank mRNA | D49915.1 | - | 2 - 3 | 345 | 104 | |
More Information
Set Membership: |
|

CHST3
Genomic Map