Assay Details
| Gene Symbol: PPP2CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205124.1 | NP_990455.1 | 5 - 6 | 820 | 53 | |
| GenBank mRNA | D17531.1 | - | 5 - 6 | 820 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP2CB| Gene Symbol: PPP2CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205124.1 | NP_990455.1 | 5 - 6 | 820 | 53 | |
| GenBank mRNA | D17531.1 | - | 5 - 6 | 820 | 53 | |
Set Membership: |
|