Assay Details
| Gene Symbol: DKK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205125.1 | NP_990456.1 | 5 - 6 | 683 | 84 | |
| XM_015286337.2 | XP_015141823.1 | 5 - 6 | 696 | 84 | ||
| GenBank mRNA | CR354079.1 | - | 4 - 5 | 543 | 84 | |
| D26311.1 | - | 5 - 6 | 683 | 84 | ||
More Information
Set Membership: |
|

DKK3
Genomic Map