Assay Details
| Gene Symbol: BMPR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205132.1 | NP_990463.1 | 6 - 7 | 704 | 59 | |
| XM_015276152.2 | XP_015131638.1 | 6 - 7 | 600 | 59 | ||
| XM_015276153.2 | XP_015131639.1 | 4 - 5 | 456 | 59 | ||
| GenBank mRNA | D13432.1 | - | 6 - 7 | 704 | 59 | |
More Information
Set Membership: |
|

BMPR1B
Genomic Map