Assay Details
| Gene Symbol: SLC16A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205140.2 | NP_990471.2 | 4 - 5 | 1683 | 83 | |
| GenBank mRNA | U15685.1 | - | 4 - 5 | 1683 | 83 | |
More Information
Set Membership: |
|

SLC16A8
Genomic Map