Assay Details
| Gene Symbol: GLRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205160.1 | NP_990491.1 | 1 - 2 | 241 | 127 | |
| GenBank mRNA | Y09235.1 | - | 241 | 127 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLRX| Gene Symbol: GLRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205160.1 | NP_990491.1 | 1 - 2 | 241 | 127 | |
| GenBank mRNA | Y09235.1 | - | 241 | 127 | ||
Set Membership: |
|