Assay Details
| Gene Symbol: EFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205184.2 | NP_990515.1 | 2 - 3 | 445 | 67 | |
| XM_015280412.2 | XP_015135898.1 | 2 - 3 | 1324 | 67 | ||
| XM_015280413.2 | XP_015135899.1 | 2 - 3 | 359 | 67 | ||
| GenBank mRNA | X90377.1 | - | 446 | 67 | ||
More Information
Set Membership: |
|

EFNA5
Genomic Map