Assay Details
| Gene Symbol: AVP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205185.2 | NP_990516.1 | 1 - 2 | 181 | 66 | |
| GenBank mRNA | CR390522.1 | - | 1 - 2 | 191 | 66 | |
| X55130.1 | - | 163 | 66 | |||
More Information
Set Membership: |
|

AVP
Genomic Map