Assay Details
| Gene Symbol: AVP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205185.2 | NP_990516.1 | 2 - 3 | 379 | 151 | |
| GenBank mRNA | CR390522.1 | - | 2 - 3 | 389 | 151 | |
| X55130.1 | - | 361 | 151 | |||
More Information
Set Membership: |
|

AVP
Genomic Map