Assay Details
| Gene Symbol: SOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205188.2 | NP_990519.2 | 1 | 961 | 78 | |
| GenBank mRNA | D50603.1 | - | 1 | 961 | 78 | |
| U12532.1 | - | 1 | 1155 | 78 | ||
More Information
Set Membership: |
|

SOX2
Genomic Map