Assay Details
| Gene Symbol: FST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205200.1 | NP_990531.1 | 2 - 3 | 275 | 57 | |
| XM_015277250.2 | XP_015132736.1 | 2 - 3 | 419 | 57 | ||
| GenBank mRNA | X87609.1 | - | 275 | 57 | ||
More Information
Set Membership: |
|

FST
Genomic Map