Assay Details
| Gene Symbol: NR1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205205.2 | NP_990536.2 | 1 - 2 | 406 | 72 | |
| XM_015281306.2 | XP_015136792.1 | 2 - 3 | 409 | 72 | ||
| XM_015281307.2 | XP_015136793.1 | 2 - 3 | 409 | 72 | ||
| GenBank mRNA | CR382426.1 | - | 1 - 2 | 406 | 72 | |
| X80258.1 | - | 336 | 72 | |||
More Information
Set Membership: |
|

NR1D2
Genomic Map