Assay Details
| Gene Symbol: MELTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205207.2 | NP_990538.2 | 8 - 9 | 1252 | 52 | |
| GenBank mRNA | X91908.1 | - | 1252 | 52 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MELTF| Gene Symbol: MELTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205207.2 | NP_990538.2 | 8 - 9 | 1252 | 52 | |
| GenBank mRNA | X91908.1 | - | 1252 | 52 | ||
Set Membership: |
|