Assay Details
| Gene Symbol: RAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205258.2 | NP_990589.1 | 2 - 3 | 151 | 67 | |
| GenBank mRNA | X66906.1 | - | 48 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAN| Gene Symbol: RAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205258.2 | NP_990589.1 | 2 - 3 | 151 | 67 | |
| GenBank mRNA | X66906.1 | - | 48 | 67 | ||
Set Membership: |
|