Assay Details
| Gene Symbol: NCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205265.1 | NP_990596.1 | 12 - 13 | 1748 | 72 | |
| GenBank mRNA | M21791.1 | - | 3 - 4 | 430 | 72 | |
| X17199.1 | - | 1748 | 72 | |||
More Information
Set Membership: |
|

NCL
Genomic Map