Assay Details
| Gene Symbol: NCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205265.1 | NP_990596.1 | 6 - 7 | 933 | 101 | |
| GenBank mRNA | X17199.1 | - | 933 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NCL| Gene Symbol: NCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205265.1 | NP_990596.1 | 6 - 7 | 933 | 101 | |
| GenBank mRNA | X17199.1 | - | 933 | 101 | ||
Set Membership: |
|