Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205281.1 | NP_990612.1 | 1 - 2 | 165 | 109 | |
| GenBank mRNA | CR390743.1 | - | 1 - 2 | 109 | ||
| V00428.1 | - | 165 | 109 | |||
More Information
Set Membership: |
|

LYZ
Genomic Map