Assay Details
| Gene Symbol: PROX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005616.1 | NP_001005616.1 | 1 - 2 | 1797 | 84 | |
| XM_015283620.2 | XP_015139106.1 | 2 - 3 | 2181 | 84 | ||
| XM_015283621.2 | XP_015139107.1 | 2 - 3 | 2308 | 84 | ||
| XM_015283622.2 | XP_015139108.1 | 2 - 3 | 2177 | 84 | ||
| GenBank mRNA | U46563.1 | - | 1 - 2 | 1797 | 84 | |
More Information
Set Membership: |
|

PROX1
Genomic Map