Assay Details
| Gene Symbol: AARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005836.1 | NP_001005836.1 | 16 - 17 | 2397 | 135 | |
| XM_015279131.2 | XP_015134617.1 | 16 - 17 | 2421 | 135 | ||
| XM_015279132.2 | XP_015134618.1 | 16 - 17 | 2397 | 135 | ||
| GenBank mRNA | AJ720131.1 | - | 16 - 17 | 2397 | 135 | |
More Information
Set Membership: |
|

AARS1
Genomic Map