Assay Details
| Gene Symbol: DNAJA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005841.1 | NP_001005841.1 | 6 - 7 | 854 | 87 | |
| GenBank mRNA | AJ720637.1 | - | 6 - 7 | 854 | 87 | |
More Information
Set Membership: |
|

DNAJA2
Genomic Map