Assay Details
| Gene Symbol: DHODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006131.2 | NP_001006131.1 | 3 - 4 | 604 | 63 | |
| XM_025154202.1 | XP_025009970.1 | 3 - 4 | 618 | 63 | ||
| GenBank mRNA | AJ721004.1 | - | 3 - 4 | 605 | 63 | |
More Information
Set Membership: |
|

DHODH
Genomic Map