Assay Details
| Gene Symbol: HMCES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006137.1 | NP_001006137.1 | 1 - 2 | 231 | 112 | |
| XM_015293076.2 | XP_015148562.1 | 1 - 2 | 242 | 112 | ||
| GenBank mRNA | AJ720353.1 | - | 1 - 2 | 231 | 112 | |
More Information
Set Membership: |
|

HMCES
Genomic Map