Assay Details
| Gene Symbol: RUVBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006138.1 | NP_001006138.1 | 4 - 5 | 580 | 69 | |
| XM_025154469.1 | XP_025010237.1 | 3 - 4 | 511 | 69 | ||
| GenBank mRNA | AJ720860.1 | - | 4 - 5 | 580 | 69 | |
| CR353587.1 | - | 4 - 5 | 592 | 69 | ||
More Information
Set Membership: |
|

RUVBL1
Genomic Map