Assay Details
| Gene Symbol: HARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006144.1 | NP_001006144.1 | 4 - 5 | 404 | 148 | |
| GenBank mRNA | AJ720198.1 | - | 4 - 5 | 404 | 148 | |
More Information
Set Membership: |
|

HARS1
Genomic Map