Assay Details
| Gene Symbol: IK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006145.1 | NP_001006145.1 | 18 - 19 | 1614 | 58 | |
| GenBank mRNA | AJ721112.1 | - | 18 - 19 | 1614 | 58 | |
| CR523801.1 | - | 3 - 4 | 503 | 58 | ||
More Information
Set Membership: |
|

IK
Genomic Map