Assay Details
| Gene Symbol: SKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006153.1 | NP_001006153.1 | 2 - 3 | 159 | 77 | |
| XM_025154856.1 | XP_025010624.1 | 2 - 3 | 173 | 77 | ||
| XM_025154857.1 | XP_025010625.1 | 2 - 3 | 143 | 77 | ||
| GenBank mRNA | AJ720129.1 | - | 2 - 3 | 159 | 77 | |
More Information
Set Membership: |
|

SKP1
Genomic Map