Assay Details
| Gene Symbol: COPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006163.1 | NP_001006163.1 | 5 - 6 | 486 | 67 | |
| XM_015294174.2 | XP_015149660.1 | 5 - 6 | 462 | 67 | ||
| XM_015294175.2 | XP_015149661.1 | 5 - 6 | 542 | 67 | ||
| XM_025155083.1 | XP_025010851.1 | 5 - 6 | 551 | 67 | ||
| GenBank mRNA | AJ720483.1 | - | 5 - 6 | 486 | 67 | |
More Information
Set Membership: |
|

COPS3
Genomic Map