Assay Details
| Gene Symbol: FOPNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006170.1 | NP_001006170.1 | 2 - 3 | 239 | 97 | |
| XM_025155101.1 | XP_025010869.1 | 2 - 3 | 283 | 97 | ||
| GenBank mRNA | AJ720610.1 | - | 2 - 3 | 239 | 97 | |
More Information
Set Membership: |
|

FOPNL
Genomic Map