Assay Details
| Gene Symbol: FOPNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006170.1 | NP_001006170.1 | 3 - 4 | 318 | 152 | |
| XM_015294217.2 | XP_015149703.1 | 3 - 4 | 359 | 152 | ||
| GenBank mRNA | AJ720610.1 | - | 3 - 4 | 318 | 152 | |
More Information
Set Membership: |
|

FOPNL
Genomic Map