Assay Details
| Gene Symbol: SEPT2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006182.2 | NP_001006182.1 | 11 - 12 | 1124 | 105 | |
| XM_015275238.2 | XP_015130724.1 | 11 - 12 | 1311 | 105 | ||
| XM_015275239.2 | XP_015130725.1 | 11 - 12 | 1126 | 105 | ||
| XM_025155432.1 | XP_025011200.1 | 10 - 11 | 1020 | 105 | ||
| XM_025155433.1 | XP_025011201.1 | 11 - 12 | 1063 | 105 | ||
| GenBank mRNA | AJ719413.1 | - | 11 - 12 | 1126 | 105 | |
More Information
Set Membership: |
|

SEPT2L
Genomic Map