Assay Details
| Gene Symbol: DDX55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006185.1 | NP_001006185.1 | 7 - 8 | 767 | 74 | |
| GenBank mRNA | AJ719696.1 | - | 7 - 8 | 767 | 74 | |
More Information
Set Membership: |
|

DDX55
Genomic Map