Assay Details
| Gene Symbol: PSMD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006189.1 | NP_001006189.1 | 3 - 4 | 445 | 71 | |
| GenBank mRNA | AJ720363.1 | - | 3 - 4 | 445 | 71 | |
| BX936274.1 | - | 3 - 4 | 402 | 71 | ||
More Information
Set Membership: |
|

PSMD9
Genomic Map