Assay Details
| Gene Symbol: PPP1CC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006190.1 | NP_001006190.1 | 6 - 7 | 1075 | 71 | |
| GenBank mRNA | AJ719895.1 | - | 6 - 7 | 1075 | 71 | |
More Information
Set Membership: |
|

PPP1CC
Genomic Map