Assay Details
| Gene Symbol: ACADS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006193.1 | NP_001006193.1 | 6 - 7 | 831 | 62 | |
| XM_015275330.2 | XP_015130816.1 | 6 - 7 | 874 | 62 | ||
| GenBank mRNA | AJ719878.1 | - | 6 - 7 | 831 | 62 | |
More Information
Set Membership: |
|

ACADS
Genomic Map