Assay Details
Gene Symbol: TRIM39.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006196.3 | NP_001006196.3 | 5 - 6 | 899 | 124 | |
GenBank mRNA | AJ720787.1 | - | 5 - 6 | 899 | 124 | |
AJ723433.1 | - | 4 - 5 | 471 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
