Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006209.1 | NP_001006209.1 | 4 - 5 | 413 | 60 | |
| XM_015279891.2 | XP_015135377.1 | 4 - 5 | 450 | 60 | ||
| GenBank mRNA | AJ719475.1 | - | 4 - 5 | 413 | 60 | |
More Information
Set Membership: |
|

KPNA2
Genomic Map