Assay Details
| Gene Symbol: PSMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006225.1 | NP_001006225.1 | 4 - 5 | 326 | 79 | |
| GenBank mRNA | AJ719466.1 | - | 4 - 5 | 326 | 79 | |
More Information
Set Membership: |
|

PSMC2
Genomic Map