Assay Details
| Gene Symbol: RPAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006231.1 | NP_001006231.1 | 6 - 7 | 732 | 75 | |
| XM_015281393.2 | XP_015136879.1 | 5 - 6 | 587 | 75 | ||
| GenBank mRNA | AJ720031.1 | - | 6 - 7 | 732 | 75 | |
More Information
Set Membership: |
|

RPAP3
Genomic Map