Assay Details
| Gene Symbol: FKBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006250.1 | NP_001006250.1 | 9 - 10 | 1408 | 61 | |
| GenBank mRNA | AJ720350.1 | - | 9 - 10 | 1408 | 61 | |
More Information
Set Membership: |
|

FKBP4
Genomic Map