Assay Details
| Gene Symbol: PDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006259.1 | NP_001006259.1 | 9 - 10 | 1073 | 65 | |
| GenBank mRNA | AJ719652.1 | - | 9 - 10 | 1073 | 65 | |
More Information
Set Membership: |
|

PDK3
Genomic Map