Assay Details
| Gene Symbol: UBAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006268.1 | NP_001006268.1 | 3 - 4 | 382 | 69 | |
| XM_015274298.2 | XP_015129784.2 | 3 - 4 | 317 | 69 | ||
| XM_025150130.1 | XP_025005898.1 | 3 - 4 | 400 | 69 | ||
| GenBank mRNA | AJ720376.1 | - | 3 - 4 | 382 | 69 | |
More Information
Set Membership: |
|

UBAC2
Genomic Map