Assay Details
| Gene Symbol: CAB39L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006272.2 | NP_001006272.2 | 4 - 5 | 624 | 90 | |
| XM_015275623.2 | XP_015131109.1 | 2 - 3 | 171 | 90 | ||
| XM_015275629.2 | XP_015131115.1 | 2 - 3 | 573 | 90 | ||
| XM_015275633.2 | XP_015131119.1 | 3 - 4 | 658 | 90 | ||
| GenBank mRNA | AJ456284.1 | - | 4 - 5 | 625 | 90 | |
| AJ719912.1 | - | 4 - 5 | 624 | 90 | ||
| CN233866.1 | - | 2 - 3 | 345 | 90 | ||
More Information
Set Membership: |
|

CAB39L
Genomic Map