Assay Details
Gene Symbol: TMEM123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006279.2 | NP_001006279.2 | 5 - 6 | 610 | 97 | |
GenBank mRNA | AJ720211.1 | - | 5 - 6 | 572 | 97 | |
AJ720988.1 | - | 5 - 6 | 604 | 98 | ||
BX933342.2 | - | 5 - 6 | 535 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
