Assay Details
| Gene Symbol: RTFDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006297.2 | NP_001006297.2 | 8 - 9 | 791 | 87 | |
| GenBank mRNA | AJ720403.1 | - | 8 - 9 | 794 | 87 | |
More Information
Set Membership: |
|

RTFDC1
Genomic Map