Assay Details
| Gene Symbol: SDF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006302.1 | NP_001006302.1 | 2 - 3 | 467 | 101 | |
| GenBank mRNA | AJ720139.1 | - | 2 - 3 | 467 | 101 | |
More Information
Set Membership: |
|

SDF4
Genomic Map