Assay Details
| Gene Symbol: NUDT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006490.2 | NP_001006490.1 | 3 - 4 | 215 | 66 | |
| XM_015287491.2 | XP_015142977.1 | 3 - 4 | 242 | 66 | ||
| XM_015287492.2 | XP_015142978.1 | 4 - 5 | 287 | 66 | ||
| XM_025150798.1 | XP_025006566.1 | 6 - 7 | 395 | 66 | ||
| GenBank mRNA | AJ720567.1 | - | 3 - 4 | 216 | 66 | |
| CR389417.1 | - | 3 - 4 | 224 | 66 | ||
More Information
Set Membership: |
|

NUDT14
Genomic Map