Assay Details
| Gene Symbol: PPP2R2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006507.1 | NP_001006507.1 | 9 - 10 | 1160 | 131 | |
| XM_015288989.2 | XP_015144475.1 | 6 - 7 | 864 | 131 | ||
| XM_015288990.1 | XP_015144476.1 | 6 - 7 | 804 | 131 | ||
| GenBank mRNA | AJ720649.1 | - | 9 - 10 | 1160 | 131 | |
More Information
Set Membership: |
|

PPP2R2D
Genomic Map