Assay Details
| Gene Symbol: ACADL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006511.2 | NP_001006511.1 | 6 - 7 | 778 | 73 | |
| GenBank mRNA | AJ720541.1 | - | 6 - 7 | 779 | 73 | |
More Information
Set Membership: |
|

ACADL
Genomic Map