Assay Details
| Gene Symbol: CNOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006521.2 | NP_001006521.2 | 6 - 7 | 710 | 71 | |
| XM_015289800.2 | XP_015145286.1 | 6 - 7 | 724 | 71 | ||
| GenBank mRNA | AJ720224.1 | - | 6 - 7 | 710 | 71 | |
More Information
Set Membership: |
|

CNOT9
Genomic Map