Assay Details
| Gene Symbol: SEC22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006526.1 | NP_001006526.1 | 4 - 5 | 801 | 70 | |
| XM_025152207.1 | XP_025007975.1 | 5 - 6 | 849 | 70 | ||
| XM_025152208.1 | XP_025007976.1 | 5 - 6 | 849 | 70 | ||
| GenBank mRNA | AJ720859.1 | - | 5 - 6 | 836 | 70 | |
More Information
Set Membership: |
|

SEC22A
Genomic Map